Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATTGTATTCCCCTCTTTTTCCTAA[A/C]CTAAGTCTCCCCAAATCCTTCAAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605693 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RAB30 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RAB30 - RAB30, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286059.1 | 5235 | UTR 3 | NP_001272988.1 | |||
NM_001286060.1 | 5235 | UTR 3 | NP_001272989.1 | |||
NM_001286061.1 | 5235 | UTR 3 | NP_001272990.1 | |||
NM_014488.4 | 5235 | UTR 3 | NP_055303.2 |