Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCCCGTCTTGCCGTAGTTGCGCAG[C/G]GCGCGCGCCATCTTCTGGTAGGTCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600958 MIM: 165170 | ||||||||||||||||||||
Literature Links: |
MYBPC3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MYBPC3 - myosin binding protein C, cardiac | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SPI1 - Spi-1 proto-oncogene | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080547.1 | 2364 | Silent Mutation | GCC,GCG | A,A 232 | NP_001074016.1 | |
NM_003120.2 | 2364 | Silent Mutation | GCC,GCG | A,A 231 | NP_003111.2 | |
XM_011520307.1 | 2364 | Silent Mutation | GCC,GCG | A,A 144 | XP_011518609.1 | |
XM_017018173.1 | 2364 | Silent Mutation | GCC,GCG | A,A 144 | XP_016873662.1 |