Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCCTTTCAAGGTCATGATGTTGGA[G/T]TCTGTTGCCTGGGTACCACCAGAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605628 | ||||||||||||||||||||
Literature Links: |
HTATIP2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HTATIP2 - HIV-1 Tat interactive protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098520.1 | 331 | Missense Mutation | GTC,TTC | V,F 123 | NP_001091990.1 | |
NM_001098521.1 | 331 | Missense Mutation | GTC,TTC | V,F 89 | NP_001091991.1 | |
NM_001098522.1 | 331 | Missense Mutation | GTC,TTC | V,F 89 | NP_001091992.1 | |
NM_001098523.1 | 331 | Missense Mutation | GTC,TTC | V,F 89 | NP_001091993.1 | |
NM_006410.4 | 331 | Missense Mutation | GTC,TTC | V,F 89 | NP_006401.3 |