Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACAGGGGCTGGGGCTGACGTTGGG[A/G]CTGAGGCTGGGGGATAGAAGCCTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602013 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
POLR2G PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
POLR2G - RNA polymerase II subunit G | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZBTB3 - zinc finger and BTB domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024784.3 | 924 | Intron | NP_079060.1 | |||
XM_011545256.2 | 924 | Missense Mutation | GCC,GTC | A,V 277 | XP_011543558.1 |