Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTCGCGACCGTGACTACGGGGATC[A/G]TCTGAGCAGAGGCTCCCATCGAGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609665 MIM: 605444 | ||||||||||||||||||||
Literature Links: |
NLRP14 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NLRP14 - NLR family pyrin domain containing 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_176822.3 | 1005 | Intron | NP_789792.1 | |||
XM_011520044.1 | 1005 | Intron | XP_011518346.1 |
RBMXL2 - RNA binding motif protein, X-linked like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014469.4 | 1005 | Missense Mutation | CAT,CGT | H,R 273 | NP_055284.3 |