Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGCGGGCAGGATCTCCGTGCCAC[A/G]ATGCAGAGGAAGGGTGAGCCCCATG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610509 MIM: 601197 | ||||||||||||||||||||
Literature Links: |
RIC3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RIC3 - RIC3 acetylcholine receptor chaperone | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135109.2 | 748 | Intron | NP_001128581.1 | |||
NM_001206671.2 | 748 | Intron | NP_001193600.1 | |||
NM_001206672.2 | 748 | Intron | NP_001193601.1 | |||
NM_024557.4 | 748 | Intron | NP_078833.3 | |||
XM_006718317.3 | 748 | Intron | XP_006718380.1 | |||
XM_006718318.3 | 748 | Intron | XP_006718381.1 | |||
XM_006718321.3 | 748 | Intron | XP_006718384.1 | |||
XM_017018287.1 | 748 | Intron | XP_016873776.1 | |||
XM_017018288.1 | 748 | Intron | XP_016873777.1 | |||
XM_017018289.1 | 748 | Intron | XP_016873778.1 | |||
XM_017018290.1 | 748 | Intron | XP_016873779.1 |
TUB - tubby bipartite transcription factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003320.4 | 748 | Silent Mutation | ACA,ACG | T,T 239 | NP_003311.2 | |
NM_177972.2 | 748 | Silent Mutation | ACA,ACG | T,T 184 | NP_813977.1 | |
XM_005253109.3 | 748 | Silent Mutation | ACA,ACG | T,T 226 | XP_005253166.1 | |
XM_011520344.2 | 748 | Silent Mutation | ACA,ACG | T,T 196 | XP_011518646.1 |