Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTGGTGCTCTCTTGTTCTGAGGG[C/G]AGGACCCTAGGCTGGCTGTCACGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608522 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RSF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RSF1 - remodeling and spacing factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016578.3 | 3884 | Silent Mutation | CTC,CTG | L,L 1330 | NP_057662.3 | |
XM_005274051.2 | 3884 | Silent Mutation | CTC,CTG | L,L 1327 | XP_005274108.1 | |
XM_017017923.1 | 3884 | Silent Mutation | CTC,CTG | L,L 1289 | XP_016873412.1 |