Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCGTGGACGTCGTGGGCAACCTCC[G/T]GGTGATCCTCTCCGTGCTCAGGAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600804 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MTNR1B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MTNR1B - melatonin receptor 1B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005959.3 | 282 | Missense Mutation | CGG,CTG | R,L 60 | NP_005950.1 | |
XM_011542839.2 | 282 | Missense Mutation | CGG,CTG | R,L 60 | XP_011541141.1 | |
XM_017017777.1 | 282 | Intron | XP_016873266.1 |