Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTACAGTGCAAAGACACCCTGTCC[G/T]TACGGGTAAGGGCAAGCTCCCAGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604641 | ||||||||||||||||||||
Literature Links: |
C11orf94 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C11orf94 - chromosome 11 open reading frame 94 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MAPK8IP1 - mitogen-activated protein kinase 8 interacting protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005456.3 | 572 | Missense Mutation | GTA,TTA | V,L 68 | NP_005447.1 | |
XM_005253226.3 | 572 | Missense Mutation | GTA,TTA | V,L 58 | XP_005253283.1 |