Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTGGAGATTCCAGATCCTTCAGA[C/G]TGTTTTTCTCTCAATCTTTGCTGTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607579 | ||||||||||||||||||||
Literature Links: |
SLC22A9 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SLC22A9 - solute carrier family 22 member 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_080866.2 | 330 | Missense Mutation | ACT,AGT | T,S 21 | NP_543142.2 | |
XM_017017159.1 | 330 | Missense Mutation | ACT,AGT | T,S 21 | XP_016872648.1 | |
XM_017017160.1 | 330 | UTR 5 | XP_016872649.1 |