Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGGTCCAGCACCTACCTGATGAC[C/T]GTGGGGTATAATTCACTTGTGTAGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607582 MIM: 607581 | ||||||||||||||||||||
Literature Links: |
SLC22A6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SLC22A6 - solute carrier family 22 member 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC22A8 - solute carrier family 22 member 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001184732.1 | 1459 | Silent Mutation | ACA,ACG | T,T 439 | NP_001171661.1 | |
NM_001184733.1 | 1459 | Silent Mutation | ACA,ACG | T,T 348 | NP_001171662.1 | |
NM_001184736.1 | 1459 | Silent Mutation | ACA,ACG | T,T 316 | NP_001171665.1 | |
NM_004254.3 | 1459 | Silent Mutation | ACA,ACG | T,T 439 | NP_004245.2 | |
XM_011545364.1 | 1459 | Silent Mutation | ACA,ACG | T,T 316 | XP_011543666.1 |