Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCCAGTGTTTCTTTGCTCAGCTCC[A/G]CGGTGACAGCTGAGATGGAGGGTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612299 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
COMMD9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
COMMD9 - COMM domain containing 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001101653.1 | 514 | Missense Mutation | GCG,GTG | A,V 129 | NP_001095123.1 | |
NM_001307932.1 | 514 | Silent Mutation | CGC,CGT | R,R 159 | NP_001294861.1 | |
NM_001307937.1 | 514 | Missense Mutation | GCG,GTG | A,V 162 | NP_001294866.1 | |
NM_014186.3 | 514 | Missense Mutation | GCG,GTG | A,V 171 | NP_054905.2 | |
XM_017017625.1 | 514 | Silent Mutation | CGC,CGT | R,R 136 | XP_016873114.1 |