Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGTCAGGCAGCTGCAGCTTCTTC[A/G]CTCAACTCCTATGGGGCTCAGGGTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612409 | ||||||||||||||||||||
Literature Links: |
RBM14 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RBM14 - RNA binding motif protein 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198836.1 | 1120 | Intron | NP_001185765.1 | |||
NM_001198837.1 | 1120 | Intron | NP_001185766.1 | |||
NM_006328.3 | 1120 | Silent Mutation | TCA,TCG | S,S 327 | NP_006319.1 |
RBM14-RBM4 - RBM14-RBM4 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198845.1 | 1120 | Intron | NP_001185774.1 | |||
NM_001198846.1 | 1120 | Intron | NP_001185775.1 |