Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTATATTTGCACTTAATTACATCG[G/T]TTTCTCCCAAAGACTTCCCCTTGTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606502 | ||||||||||||||||||||
Literature Links: |
MS4A14 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MS4A14 - membrane spanning 4-domains A14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001079692.2 | 771 | Missense Mutation | GGT,GTT | G,V 69 | NP_001073160.1 | |
NM_001261827.1 | 771 | Missense Mutation | GGT,GTT | G,V 69 | NP_001248756.1 | |
NM_001261828.1 | 771 | Missense Mutation | GGT,GTT | G,V 69 | NP_001248757.1 | |
NM_032597.4 | 771 | Missense Mutation | GGT,GTT | G,V 69 | NP_115986.3 |
MS4A7 - membrane spanning 4-domains A7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |