Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATGACCCATTCCTGCTGCTGGAAG[A/G]CTAACTGGTATTGTCTGATGTCCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615265 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AGBL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AGBL2 - ATP/GTP binding protein like 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FNBP4 - formin binding protein 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318339.1 | 2719 | Silent Mutation | AGC,AGT | S,S 845 | NP_001305268.1 | |
NM_015308.3 | 2719 | Silent Mutation | AGC,AGT | S,S 843 | NP_056123.2 |