Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGTTCACTGTGCGCCCGTCAACTG[C/T]GCTCTGCGCGCTGTAATTGTCGAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615187 MIM: 179505 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PGAP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PGAP2 - post-GPI attachment to proteins 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RHOG - ras homolog family member G | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001665.3 | 471 | Missense Mutation | ACA,GCA | T,A 45 | NP_001656.2 | |
XM_005252916.2 | 471 | Missense Mutation | ACA,GCA | T,A 45 | XP_005252973.1 | |
XM_017017719.1 | 471 | Missense Mutation | ACA,GCA | T,A 45 | XP_016873208.1 |