Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCAGGTGCCTTGGCGGTCACTTC[A/G]CACTGTCACATTGAAGGCTAGAAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609651 MIM: 601644 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPHA2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GPHA2 - glycoprotein hormone alpha 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_130769.3 | 921 | Nonsense Mutation | CGA,TGA | R,* 41 | NP_570125.1 | |
XM_011544774.2 | 921 | Nonsense Mutation | CGA,TGA | R,* 74 | XP_011543076.1 | |
XM_011544775.2 | 921 | Nonsense Mutation | CGA,TGA | R,* 41 | XP_011543077.1 | |
XM_011544776.2 | 921 | Nonsense Mutation | CGA,TGA | R,* 41 | XP_011543078.1 |
MAJIN - membrane anchored junction protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PPP2R5B - protein phosphatase 2 regulatory subunit B'beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006244.3 | 921 | Intron | NP_006235.1 | |||
XM_011545132.2 | 921 | Intron | XP_011543434.1 | |||
XM_011545133.1 | 921 | Intron | XP_011543435.1 | |||
XM_011545134.2 | 921 | Intron | XP_011543436.1 |