Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGATGAGGATGCAGAGCGCCGCCGC[C/T]GCCACAATGCCCACCACCATGCCCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600566 MIM: 607096 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NRXN2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NRXN2 - neurexin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015080.3 | 3292 | Silent Mutation | GCA,GCG | A,A 1644 | NP_055895.1 | |
NM_138732.2 | 3292 | Silent Mutation | GCA,GCG | A,A 1574 | NP_620060.1 | |
NM_138734.2 | 3292 | Silent Mutation | GCA,GCG | A,A 598 | NP_620063.1 | |
XM_005274400.3 | 3292 | Silent Mutation | GCA,GCG | A,A 1637 | XP_005274457.1 | |
XM_005274401.3 | 3292 | Silent Mutation | GCA,GCG | A,A 1443 | XP_005274458.1 | |
XM_005274402.3 | 3292 | Silent Mutation | GCA,GCG | A,A 1435 | XP_005274459.1 | |
XM_011545370.1 | 3292 | Silent Mutation | GCA,GCG | A,A 1647 | XP_011543672.1 | |
XM_011545371.1 | 3292 | Silent Mutation | GCA,GCG | A,A 1646 | XP_011543673.1 | |
XM_011545373.1 | 3292 | Silent Mutation | GCA,GCG | A,A 1640 | XP_011543675.1 | |
XM_011545375.1 | 3292 | Silent Mutation | GCA,GCG | A,A 1632 | XP_011543677.1 | |
XM_011545385.1 | 3292 | Silent Mutation | GCA,GCG | A,A 909 | XP_011543687.1 | |
XM_017018563.1 | 3292 | Silent Mutation | GCA,GCG | A,A 1637 | XP_016874052.1 | |
XM_017018564.1 | 3292 | Silent Mutation | GCA,GCG | A,A 1636 | XP_016874053.1 | |
XM_017018565.1 | 3292 | Silent Mutation | GCA,GCG | A,A 1635 | XP_016874054.1 | |
XM_017018566.1 | 3292 | Silent Mutation | GCA,GCG | A,A 1623 | XP_016874055.1 | |
XM_017018567.1 | 3292 | Silent Mutation | GCA,GCG | A,A 1617 | XP_016874056.1 | |
XM_017018568.1 | 3292 | Silent Mutation | GCA,GCG | A,A 1614 | XP_016874057.1 | |
XM_017018569.1 | 3292 | Silent Mutation | GCA,GCG | A,A 1613 | XP_016874058.1 | |
XM_017018570.1 | 3292 | Silent Mutation | GCA,GCG | A,A 1453 | XP_016874059.1 | |
XM_017018571.1 | 3292 | Silent Mutation | GCA,GCG | A,A 1450 | XP_016874060.1 | |
XM_017018572.1 | 3292 | Silent Mutation | GCA,GCG | A,A 1412 | XP_016874061.1 | |
XM_017018573.1 | 3292 | Silent Mutation | GCA,GCG | A,A 1411 | XP_016874062.1 | |
XM_017018574.1 | 3292 | Silent Mutation | GCA,GCG | A,A 1402 | XP_016874063.1 | |
XM_017018575.1 | 3292 | Silent Mutation | GCA,GCG | A,A 1401 | XP_016874064.1 | |
XM_017018576.1 | 3292 | Silent Mutation | GCA,GCG | A,A 1228 | XP_016874065.1 | |
XM_017018577.1 | 3292 | Silent Mutation | GCA,GCG | A,A 906 | XP_016874066.1 | |
XM_017018578.1 | 3292 | Silent Mutation | GCA,GCG | A,A 1420 | XP_016874067.1 |
SLC22A12 - solute carrier family 22 member 12 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |