Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTACCTAGGAAGGAAGTACTATTTG[A/C]GAATCTTGTGTAGTAAGGTGATTTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609490 | ||||||||||||||||||||
Literature Links: |
CARD17 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CARD17 - caspase recruitment domain family member 17 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001007232.1 | 329 | Silent Mutation | TCG,TCT | S,S 104 | NP_001007233.1 | |
XM_011542838.2 | 329 | Silent Mutation | TCG,TCT | S,S 119 | XP_011541140.1 |