Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCATCGAGGCAGAGGCTCTATGGCT[C/T]CTCCCCCATCTGCAGCAGGGAGTTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606277 | ||||||||||||||||||||
Literature Links: |
TOLLIP PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TOLLIP - toll interacting protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318512.1 | 981 | Missense Mutation | AAG,GAG | K,E 223 | NP_001305441.1 | |
NM_001318514.1 | 981 | Missense Mutation | AAG,GAG | K,E 204 | NP_001305443.1 | |
NM_001318515.1 | 981 | UTR 3 | NP_001305444.1 | |||
NM_001318516.1 | 981 | Missense Mutation | AAG,GAG | K,E 212 | NP_001305445.1 | |
NM_019009.3 | 981 | Missense Mutation | AAG,GAG | K,E 273 | NP_061882.2 | |
XM_017017931.1 | 981 | Missense Mutation | AAG,GAG | K,E 147 | XP_016873420.1 |