Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAAGCTTCCGTCAGGAGGGTGCCA[A/G]GCAGAGGAGGCCACCTGGACACTAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 169730 MIM: 611115 | ||||||||||||||||||||
Literature Links: |
PGA5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PGA5 - pepsinogen 5, group I (pepsinogen A) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VWCE - von Willebrand factor C and EGF domains | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152718.2 | 2544 | Missense Mutation | CCT,CTT | P,L 891 | NP_689931.2 | |
XM_006718460.1 | 2544 | Missense Mutation | CCT,CTT | P,L 883 | XP_006718523.1 | |
XM_006718462.1 | 2544 | Missense Mutation | CCT,CTT | P,L 831 | XP_006718525.1 | |
XM_011544817.1 | 2544 | Missense Mutation | CCT,CTT | P,L 793 | XP_011543119.1 | |
XM_017017340.1 | 2544 | Missense Mutation | CCT,CTT | P,L 870 | XP_016872829.1 | |
XM_017017341.1 | 2544 | Missense Mutation | CCT,CTT | P,L 848 | XP_016872830.1 | |
XM_017017342.1 | 2544 | Missense Mutation | CCT,CTT | P,L 761 | XP_016872831.1 | |
XM_017017343.1 | 2544 | Missense Mutation | CCT,CTT | P,L 608 | XP_016872832.1 | |
XM_017017344.1 | 2544 | Intron | XP_016872833.1 |