Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTCATAATTTTTGCAACCAATGC[A/G]TTTGCAAATAGAAGAACACATAATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604374 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TESMIN PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TESMIN - testis expressed metallothionein like protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039656.1 | 1175 | Intron | NP_001034745.1 | |||
NM_004923.3 | 1175 | Missense Mutation | CGC,TGC | R,C 397 | NP_004914.2 | |
XM_011545402.1 | 1175 | Missense Mutation | CGC,TGC | R,C 397 | XP_011543704.1 | |
XM_011545403.2 | 1175 | Missense Mutation | CGC,TGC | R,C 397 | XP_011543705.1 | |
XM_017018588.1 | 1175 | Missense Mutation | CGC,TGC | R,C 331 | XP_016874077.1 |