Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTGATCTAAACGGGTAACCATCTC[C/T]TCCAGTTTCTGGTGACCCTTTTTCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601387 | ||||||||||||||||||||
Literature Links: |
LDHAL6A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LDHAL6A - lactate dehydrogenase A like 6A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TSG101 - tumor susceptibility 101 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006292.3 | 953 | Silent Mutation | GAA,GAG | E,E 271 | NP_006283.1 | |
XM_005253108.3 | 953 | Silent Mutation | GAA,GAG | E,E 219 | XP_005253165.1 |