Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAAAAGTTGTGTCCTTTGCTTCAC[C/T]GTGATCCATTGAATGATCATTCTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606548 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MS4A6A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MS4A6A - membrane spanning 4-domains A6A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001247999.1 | 1356 | Intron | NP_001234928.1 | |||
NM_022349.3 | 1356 | Intron | NP_071744.2 | |||
NM_152851.2 | 1356 | UTR 3 | NP_690590.1 | |||
NM_152852.2 | 1356 | UTR 3 | NP_690591.1 | |||
XM_005274177.2 | 1356 | Intron | XP_005274234.1 | |||
XM_006718658.2 | 1356 | Intron | XP_006718721.1 | |||
XM_006718660.2 | 1356 | Intron | XP_006718723.1 | |||
XM_006718661.2 | 1356 | Intron | XP_006718724.1 | |||
XM_011545209.1 | 1356 | Intron | XP_011543511.1 | |||
XM_017018123.1 | 1356 | Intron | XP_016873612.1 | |||
XM_017018124.1 | 1356 | Intron | XP_016873613.1 | |||
XM_017018125.1 | 1356 | Intron | XP_016873614.1 |