Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCCCTCCGCCCTCAGGCCGCTGC[C/T]CTCCGGCCTGGTGGGGGGCGCTGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602047 MIM: 602854 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PDE3B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PDE3B - phosphodiesterase 3B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000922.3 | 1128 | Missense Mutation | CCC,CTC | P,L 248 | NP_000913.2 | |
XM_006718249.3 | 1128 | Missense Mutation | CCC,CTC | P,L 248 | XP_006718312.1 | |
XM_017017911.1 | 1128 | Missense Mutation | CCC,CTC | P,L 248 | XP_016873400.1 | |
XM_017017912.1 | 1128 | Missense Mutation | CCC,CTC | P,L 248 | XP_016873401.1 |
PSMA1 - proteasome subunit alpha 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |