Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTACGTCAGGTAGAACTTGGAGAA[C/T]ACCTTGTACCCCTGGGCCTTAATGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
KCTD14 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KCTD14 - potassium channel tetramerization domain containing 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282406.1 | 1330 | Silent Mutation | GTA,GTG | V,V 198 | NP_001269335.1 | |
NM_023930.3 | 1330 | Silent Mutation | GTA,GTG | V,V 228 | NP_076419.2 |
NDUFC2-KCTD14 - NDUFC2-KCTD14 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001203260.1 | 1330 | UTR 3 | NP_001190189.1 | |||
NM_001203261.1 | 1330 | UTR 3 | NP_001190190.1 | |||
NM_001203262.1 | 1330 | UTR 3 | NP_001190191.1 |