Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCGGCCTCCAGGCCTTTTCTTTA[C/G]TTTTTCTTTGTAGGTAGGATAACCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602550 MIM: 615933 | ||||||||||||||||||||
Literature Links: |
ARNTL PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ARNTL - aryl hydrocarbon receptor nuclear translocator like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
BTBD10 - BTB domain containing 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297741.1 | 2520 | Intron | NP_001284670.1 | |||
NM_001297742.1 | 2520 | Missense Mutation | CTA,GTA | L,V 389 | NP_001284671.1 | |
NM_032320.6 | 2520 | Intron | NP_115696.2 | |||
XM_017018405.1 | 2520 | Missense Mutation | CTA,GTA | L,V 424 | XP_016873894.1 | |
XM_017018406.1 | 2520 | Intron | XP_016873895.1 | |||
XM_017018407.1 | 2520 | Missense Mutation | CTA,GTA | L,V 416 | XP_016873896.1 | |
XM_017018408.1 | 2520 | Missense Mutation | CTA,GTA | L,V 381 | XP_016873897.1 | |
XM_017018409.1 | 2520 | Intron | XP_016873898.1 | |||
XM_017018410.1 | 2520 | Intron | XP_016873899.1 | |||
XM_017018411.1 | 2520 | Intron | XP_016873900.1 |