Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCCAGAGCAAAAGGAGGATGGTG[A/C]AGAAGAGCACCCCAAACTGCAGCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606158 MIM: 608941 | ||||||||||||||||||||
Literature Links: |
BSCL2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BSCL2 - BSCL2, seipin lipid droplet biogenesis associated | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001122955.3 | 811 | Missense Mutation | TGC,TTC | C,F 100 | NP_001116427.1 | |
NM_001130702.2 | 811 | Missense Mutation | TGC,TTC | C,F 36 | NP_001124174.2 | |
NM_032667.6 | 811 | Missense Mutation | TGC,TTC | C,F 36 | NP_116056.3 |
GNG3 - G protein subunit gamma 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HNRNPUL2 - heterogeneous nuclear ribonucleoprotein U like 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HNRNPUL2-BSCL2 - HNRNPUL2-BSCL2 readthrough (NMD candidate) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |