Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTCCTCCTCCTCCATCGCCGCCT[A/C]CTATCCTGTCAGCCGCAGTCGTGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARHGEF17 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARHGEF17 - Rho guanine nucleotide exchange factor 17 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014786.3 | 737 | Missense Mutation | TAC,TCC | Y,S 239 | NP_055601.2 | |
XM_017018623.1 | 737 | Intron | XP_016874112.1 | |||
XM_017018624.1 | 737 | Intron | XP_016874113.1 |