Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAATGACACGGAGGCCACCATGGC[C/T]GATGGCGGGGCCGAGGAAGAAGAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 176797 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZBTB16 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZBTB16 - zinc finger and BTB domain containing 16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001018011.1 | 1118 | Silent Mutation | GCC,GCT | A,A 138 | NP_001018011.1 | |
NM_006006.4 | 1118 | Silent Mutation | GCC,GCT | A,A 138 | NP_005997.2 | |
XM_005271658.4 | 1118 | Silent Mutation | GCC,GCT | A,A 138 | XP_005271715.1 | |
XM_017018257.1 | 1118 | Silent Mutation | GCC,GCT | A,A 138 | XP_016873746.1 | |
XM_017018258.1 | 1118 | Silent Mutation | GCC,GCT | A,A 138 | XP_016873747.1 | |
XM_017018259.1 | 1118 | Silent Mutation | GCC,GCT | A,A 138 | XP_016873748.1 |