Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCTGCTGTTCTACCAACACCAGCC[A/G]GGAAGCCCATAAGGATGTTTCCTAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 136430 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FOLR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FOLR1 - folate receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000802.3 | 260 | Missense Mutation | CAG,CGG | Q,R 72 | NP_000793.1 | |
NM_016724.2 | 260 | Missense Mutation | CAG,CGG | Q,R 72 | NP_057936.1 | |
NM_016725.2 | 260 | Missense Mutation | CAG,CGG | Q,R 72 | NP_057937.1 | |
NM_016729.2 | 260 | Missense Mutation | CAG,CGG | Q,R 72 | NP_057941.1 |