Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGCCCGCACCAACTATGAGAAAG[C/T]CATCTCCCAGGGCCACCAGGCCTTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602929 | ||||||||||||||||||||
Literature Links: |
VWA5A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
VWA5A - von Willebrand factor A domain containing 5A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130142.1 | 478 | Missense Mutation | GCC,GTC | A,V 90 | NP_001123614.1 | |
NM_014622.4 | 478 | Missense Mutation | GCC,GTC | A,V 90 | NP_055437.2 | |
NM_198315.2 | 478 | Missense Mutation | GCC,GTC | A,V 90 | NP_938057.1 | |
XM_011542828.2 | 478 | Missense Mutation | GCC,GTC | A,V 106 | XP_011541130.1 |