Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCCTATCGGAAGGATGATTTCGTT[A/T]TGGTTAGTATGGGGGTCGGAGAGTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
C11orf70 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C11orf70 - chromosome 11 open reading frame 70 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195005.1 | 252 | Missense Mutation | ATG,TTG | M,L 64 | NP_001181934.1 | |
NM_032930.2 | 252 | Missense Mutation | ATG,TTG | M,L 64 | NP_116319.2 | |
XM_005271713.3 | 252 | Missense Mutation | ATG,TTG | M,L 64 | XP_005271770.1 | |
XM_017018454.1 | 252 | Missense Mutation | ATG,TTG | M,L 64 | XP_016873943.1 |