Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCTCCTGATGGGCTCTCAACACA[C/T]GCATGGACATGGGAACACACGCAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 176879 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IGSF22 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IGSF22 - immunoglobulin superfamily member 22 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PTPN5 - protein tyrosine phosphatase, non-receptor type 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039970.1 | 3687 | UTR 3 | NP_001035059.1 | |||
NM_001278236.1 | 3687 | UTR 3 | NP_001265165.1 | |||
NM_001278238.1 | 3687 | UTR 3 | NP_001265167.1 | |||
NM_001278239.1 | 3687 | UTR 3 | NP_001265168.1 | |||
NM_006906.1 | 3687 | UTR 3 | NP_008837.1 | |||
NM_032781.3 | 3687 | UTR 3 | NP_116170.3 | |||
XM_011520411.2 | 3687 | UTR 3 | XP_011518713.1 | |||
XM_017018434.1 | 3687 | UTR 3 | XP_016873923.1 | |||
XM_017018435.1 | 3687 | UTR 3 | XP_016873924.1 | |||
XM_017018436.1 | 3687 | UTR 3 | XP_016873925.1 | |||
XM_017018437.1 | 3687 | UTR 3 | XP_016873926.1 | |||
XM_017018438.1 | 3687 | UTR 3 | XP_016873927.1 | |||
XM_017018439.1 | 3687 | UTR 3 | XP_016873928.1 | |||
XM_017018440.1 | 3687 | UTR 3 | XP_016873929.1 | |||
XM_017018441.1 | 3687 | Intron | XP_016873930.1 |