Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACGGCCAGAGAAAAGACCTAGGCG[A/G]TCGATGTTGCGGTGGAGGACACTGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 104160 MIM: 611349 | ||||||||||||||||||||
Literature Links: |
GANAB PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GANAB - glucosidase II alpha subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278192.1 | 3044 | Intron | NP_001265121.1 | |||
NM_001278193.1 | 3044 | Intron | NP_001265122.1 | |||
NM_001278194.1 | 3044 | Intron | NP_001265123.1 | |||
NM_198334.2 | 3044 | Intron | NP_938148.1 | |||
NM_198335.3 | 3044 | Intron | NP_938149.2 | |||
XM_017017412.1 | 3044 | Intron | XP_016872901.1 | |||
XM_017017413.1 | 3044 | Intron | XP_016872902.1 | |||
XM_017017414.1 | 3044 | Intron | XP_016872903.1 |
INTS5 - integrator complex subunit 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030628.1 | 3044 | Silent Mutation | GAC,GAT | D,D 997 | NP_085131.1 |