Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATCTTGGAAGTCCTGAAAATTTCG[C/T]TGGTGGCAGGTAATGGCTGCCTTCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604374 | ||||||||||||||||||||
Literature Links: |
TESMIN PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TESMIN - testis expressed metallothionein like protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039656.1 | 1283 | Intron | NP_001034745.1 | |||
NM_004923.3 | 1283 | Missense Mutation | NP_004914.2 | |||
XM_011545402.1 | 1283 | Missense Mutation | XP_011543704.1 | |||
XM_011545403.2 | 1283 | Missense Mutation | XP_011543705.1 | |||
XM_017018588.1 | 1283 | Missense Mutation | XP_016874077.1 |