Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCCGACCTGCAGCCACTTCCACCC[A/G]GCTCTTCACTGACCCGCTGGCACTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602180 | ||||||||||||||||||||
Literature Links: |
MIR4489 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MIR4489 - microRNA 4489 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR4690 - microRNA 4690 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PCNX3 - pecanex homolog 3 (Drosophila) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SIPA1 - signal-induced proliferation-associated 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006747.3 | 467 | Missense Mutation | CAG,CGG | Q,R 87 | NP_006738.3 | |
NM_153253.29 | 467 | Missense Mutation | CAG,CGG | Q,R 87 | NP_694985.29 | |
XM_005274189.2 | 467 | Missense Mutation | CAG,CGG | Q,R 87 | XP_005274246.1 | |
XM_011545214.1 | 467 | Missense Mutation | CAG,CGG | Q,R 87 | XP_011543516.1 |