Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGGTGTGTCCTTCCTGACCTTCCA[C/T]CTCCACCAGGTGCCGACACTTCCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605625 | ||||||||||||||||||||
Literature Links: |
MYEOV PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MYEOV - myeloma overexpressed | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001293291.1 | 693 | Silent Mutation | CAC,CAT | H,H 44 | NP_001280220.1 | |
NM_001293294.1 | 693 | UTR 5 | NP_001280223.1 | |||
NM_001293296.1 | 693 | UTR 5 | NP_001280225.1 | |||
NM_001300923.1 | 693 | UTR 5 | NP_001287852.1 | |||
NM_001300924.1 | 693 | Intron | NP_001287853.1 | |||
NM_138768.3 | 693 | Silent Mutation | CAC,CAT | H,H 44 | NP_620123.2 |