Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTACACGCCTGCCTGCTCCATGCT[A/G]AAGGAGACTTCGGGGTGCTTGTAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 107680 MIM: 614776 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
APOA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
APOA1 - apolipoprotein A1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
APOA1-AS - APOA1 antisense RNA | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SIK3 - SIK family kinase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001281748.1 | 3800 | Silent Mutation | TTC,TTT | F,F 1095 | NP_001268677.1 | |
NM_001281749.1 | 3800 | Silent Mutation | TTC,TTT | F,F 1254 | NP_001268678.1 | |
NM_025164.4 | 3800 | Silent Mutation | TTC,TTT | F,F 1314 | NP_079440.3 | |
XM_005271481.2 | 3800 | Silent Mutation | TTC,TTT | F,F 1362 | XP_005271538.1 | |
XM_005271482.4 | 3800 | Silent Mutation | TTC,TTT | F,F 1314 | XP_005271539.2 | |
XM_005271484.3 | 3800 | Silent Mutation | TTC,TTT | F,F 1254 | XP_005271541.2 | |
XM_005271485.3 | 3800 | Silent Mutation | TTC,TTT | F,F 763 | XP_005271542.1 | |
XM_011542721.2 | 3800 | Silent Mutation | TTC,TTT | F,F 1362 | XP_011541023.1 | |
XM_011542722.1 | 3800 | Silent Mutation | TTC,TTT | F,F 1302 | XP_011541024.1 | |
XM_011542723.2 | 3800 | Silent Mutation | TTC,TTT | F,F 1203 | XP_011541025.1 | |
XM_011542724.2 | 3800 | Silent Mutation | TTC,TTT | F,F 1203 | XP_011541026.1 | |
XM_011542725.2 | 3800 | Silent Mutation | TTC,TTT | F,F 1203 | XP_011541027.1 | |
XM_011542726.1 | 3800 | Intron | XP_011541028.1 | |||
XM_017017424.1 | 3800 | Silent Mutation | TTC,TTT | F,F 1206 | XP_016872913.1 | |
XM_017017425.1 | 3800 | Silent Mutation | TTC,TTT | F,F 1203 | XP_016872914.1 | |
XM_017017426.1 | 3800 | Silent Mutation | TTC,TTT | F,F 1203 | XP_016872915.1 | |
XM_017017427.1 | 3800 | Silent Mutation | TTC,TTT | F,F 715 | XP_016872916.1 |