Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTGATATCCTTAGTTACTCCACC[A/T]CCTGACCAGGGGTCTAGTTCACCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 176785 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRCP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PRCP - prolylcarboxypeptidase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319214.1 | 1566 | Silent Mutation | GGA,GGT | G,G 329 | NP_001306143.1 | |
NM_005040.3 | 1566 | Silent Mutation | GGA,GGT | G,G 434 | NP_005031.1 | |
NM_199418.3 | 1566 | Silent Mutation | GGA,GGT | G,G 455 | NP_955450.2 | |
XM_005274093.1 | 1566 | Silent Mutation | GGA,GGT | G,G 329 | XP_005274150.1 |