Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCACAGGCTGAGTGGAAGAAACCTC[A/G]AACAATGGTGTGCAGGACAAAGGTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603733 | ||||||||||||||||||||
Literature Links: |
SLC43A1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SLC43A1 - solute carrier family 43 member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198810.1 | 1461 | Nonsense Mutation | CGA,TGA | R,* 456 | NP_001185739.1 | |
NM_003627.5 | 1461 | Nonsense Mutation | CGA,TGA | R,* 456 | NP_003618.1 | |
XM_005274358.4 | 1461 | Nonsense Mutation | CGA,TGA | R,* 472 | XP_005274415.1 | |
XM_011545320.2 | 1461 | Nonsense Mutation | CGA,TGA | R,* 441 | XP_011543622.1 | |
XM_011545321.2 | 1461 | Nonsense Mutation | CGA,TGA | R,* 200 | XP_011543623.1 | |
XM_011545322.1 | 1461 | Nonsense Mutation | CGA,TGA | R,* 200 | XP_011543624.1 | |
XM_017018451.1 | 1461 | Nonsense Mutation | CGA,TGA | R,* 456 | XP_016873940.1 | |
XM_017018452.1 | 1461 | Nonsense Mutation | CGA,TGA | R,* 425 | XP_016873941.1 | |
XM_017018453.1 | 1461 | Nonsense Mutation | CGA,TGA | R,* 200 | XP_016873942.1 |