Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGAGCACTCCAGTCTGAACACTT[A/T]AGTTATGGAAATTAAAAAAAAAAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608752 MIM: 606227 MIM: 604725 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C1QTNF5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C1QTNF5 - C1q and tumor necrosis factor related protein 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MFRP - membrane frizzled-related protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
USP2 - ubiquitin specific peptidase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243759.1 | 1651 | UTR 3 | NP_001230688.1 | |||
NM_004205.4 | 1651 | UTR 3 | NP_004196.4 | |||
NM_171997.2 | 1651 | UTR 3 | NP_741994.1 | |||
XM_005271721.4 | 1651 | UTR 3 | XP_005271778.1 | |||
XM_005271722.2 | 1651 | UTR 3 | XP_005271779.1 | |||
XM_017018539.1 | 1651 | UTR 3 | XP_016874028.1 |