Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCCATGCAGCTGCAGCAGCCTCA[C/T]TTGCTGCACTTCCTGGGGGAGCATT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610109 MIM: 613160 | ||||||||||||||||||||
Literature Links: |
ANO2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ANO2 - anoctamin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278596.1 | 8694 | Intron | NP_001265525.1 | |||
NM_001278597.1 | 8694 | Intron | NP_001265526.1 | |||
XM_006718988.3 | 8694 | Intron | XP_006719051.1 | |||
XM_011520975.2 | 8694 | Intron | XP_011519277.1 | |||
XM_011520978.2 | 8694 | Intron | XP_011519280.1 | |||
XM_017019672.1 | 8694 | Intron | XP_016875161.1 |
VWF - von Willebrand factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000552.4 | 8694 | Silent Mutation | NP_000543.2 |