Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACCCGGCGATATGGCTGCCGCTGT[G/T]CCCCGCGCCGCATTTCTCTCCCCGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602287 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ERP29 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ERP29 - endoplasmic reticulum protein 29 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001034025.1 | 133 | Silent Mutation | GTG,GTT | V,V 5 | NP_001029197.1 | |
NM_006817.3 | 133 | Silent Mutation | GTG,GTT | V,V 5 | NP_006808.1 | |
XM_017018720.1 | 133 | Intron | XP_016874209.1 |