Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGAAGCCAGAATGAATGGCCACAA[C/T]GCCTGATGTTAGACTTTCGTAAGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616359 MIM: 601562 MIM: 616207 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
COQ5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
COQ5 - coenzyme Q5, methyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032314.3 | 972 | Missense Mutation | ATT,GTT | I,V 318 | NP_115690.3 | |
XM_006719639.2 | 972 | Missense Mutation | ATT,GTT | I,V 237 | XP_006719702.1 |
DYNLL1 - dynein light chain LC8-type 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NRAV - negative regulator of antiviral response (non-protein coding) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |