Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTCTAGACTTTACTCATCTATGT[G/T]CTCTTTCTTTGCACTGTAAATATTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609095 | ||||||||||||||||||||
Literature Links: |
FBXO21 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FBXO21 - F-box protein 21 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015002.2 | 1737 | Missense Mutation | AAC,CAC | N,H 618 | NP_055817.1 | |
NM_033624.2 | 1737 | Missense Mutation | AAC,CAC | N,H 625 | NP_296373.1 | |
XM_017019037.1 | 1737 | Missense Mutation | AAC,CAC | N,H 565 | XP_016874526.1 | |
XM_017019038.1 | 1737 | Missense Mutation | AAC,CAC | N,H 558 | XP_016874527.1 | |
XM_017019039.1 | 1737 | Missense Mutation | AAC,CAC | N,H 465 | XP_016874528.1 |
TESC-AS1 - TESC antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |