Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACGAACAAAGTCACTCGGATCCAGA[A/G]CATGAATTATGGCACCATTAAGTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602566 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
P2RX7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
P2RX7 - purinergic receptor P2X 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002562.5 | 220 | Missense Mutation | AAC,AGC | N,S 23 | NP_002553.3 | |
XM_011538419.2 | 220 | UTR 5 | XP_011536721.1 | |||
XM_011538420.2 | 220 | UTR 5 | XP_011536722.1 | |||
XM_017019364.1 | 220 | UTR 5 | XP_016874853.1 | |||
XM_017019365.1 | 220 | UTR 5 | XP_016874854.1 | |||
XM_017019366.1 | 220 | UTR 5 | XP_016874855.1 | |||
XM_017019367.1 | 220 | UTR 5 | XP_016874856.1 |