Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGTCAGGAGACAGCGCAGCTGGTG[A/G]GCTCCACTGCCGTGAGTGCTTCACC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605571 | ||||||||||||||||||||
Literature Links: |
PIWIL1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PIWIL1 - piwi like RNA-mediated gene silencing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001190971.1 | 281 | Missense Mutation | AGC,GGC | S,G 23 | NP_001177900.1 | |
NM_004764.4 | 281 | Missense Mutation | AGC,GGC | S,G 23 | NP_004755.2 | |
XM_011539002.2 | 281 | Missense Mutation | AGC,GGC | S,G 23 | XP_011537304.1 | |
XM_011539003.2 | 281 | Missense Mutation | AGC,GGC | S,G 23 | XP_011537305.1 | |
XM_011539004.2 | 281 | Missense Mutation | AGC,GGC | S,G 23 | XP_011537306.1 | |
XM_011539005.1 | 281 | Missense Mutation | AGC,GGC | S,G 23 | XP_011537307.1 | |
XM_011539006.2 | 281 | Intron | XP_011537308.1 | |||
XM_017020229.1 | 281 | Missense Mutation | AGC,GGC | S,G 23 | XP_016875718.1 |