Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCATGTATTCTTGGTCCCAGACTT[C/T]GAAAGTACACATGTTTCATGGCCTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603440 | ||||||||||||||||||||
Literature Links: |
CDK17 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CDK17 - cyclin dependent kinase 17 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001170464.2 | 2053 | Missense Mutation | CAA,CGA | Q,R 474 | NP_001163935.1 | |
NM_002595.4 | 2053 | Missense Mutation | CAA,CGA | Q,R 474 | NP_002586.2 | |
XM_017019405.1 | 2053 | Missense Mutation | CAA,CGA | Q,R 474 | XP_016874894.1 | |
XM_017019406.1 | 2053 | Missense Mutation | CAA,CGA | Q,R 474 | XP_016874895.1 | |
XM_017019407.1 | 2053 | Missense Mutation | CAA,CGA | Q,R 453 | XP_016874896.1 |