Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGCCATGAAGCCAGCGCTGCTGGA[A/G]GTGATGAGGATGAACAGAATCTGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610128 | ||||||||||||||||||||
Literature Links: |
CHST11 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CHST11 - carbohydrate (chondroitin 4) sulfotransferase 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001173982.1 | 519 | Silent Mutation | GAA,GAG | E,E 7 | NP_001167453.1 | |
NM_018413.5 | 519 | Silent Mutation | GAA,GAG | E,E 7 | NP_060883.1 | |
XM_017019369.1 | 519 | Intron | XP_016874858.1 |